Learn More
thrombospondin 4, Mouse, Purified MaxPab™ Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a full-length human THBS4 protein.
Supplier: Abnova Corporation H00007060B01P
Description
The protein encoded by this gene belongs to the thrombospondin protein family. Thrombospondin family members are adhesive glycoproteins that mediate cell-to-cell and cell-to-matrix interactions. This protein forms a pentamer and can bind to heparin and calcium. Studies of the rat counterpart suggest that this protein may be involved in local signaling in the developing and adult nervous system. [provided by RefSeq
Sequence: MLAPRGAAVLLLHLVLQRWLAAGAQATPQVFDLLPSSSQRLNPGALLPVLTDPALNDLYVISTFKLQTKSSATIFGLYSSTDNSKYFEFTVMGRLNKAILRYLKNDGKVHLVVFNNLQLADGRRHRILLRLSNLQRGAGSLELYLDCIQVDSVHNLPRAFAGPSQKPETIELRTFQRKPQDFLEELKLVVRGSLFQVASLQDCFLQQSEPLAATGTGDFNRQFLGQMTQLNQLLGEVKDLLRQQVKETSFLRNTIAECQACGPLKFQSPTPSTVVPPAPPAPPTRPPRRCDSNPCFRGVQCTDSRDGFQCGPCPEGYTGNGITCIDVDECKYHPCYPGVHCINLSPGFRCDACPVGFTGPMVQGVGISFAKSNKQVCTDIDECRNGACVPNSICVNTLGSYRCGPCKPGYTGDQIRGCKAERNCRNPELNPCSVNAQCIEERQGDVTCVCGVGWAGDGYICGKDVDIDSYPDEELPCSARNCKKDNCKYVPNSGQEDADRDGIGDACDEDADGDGILNEQDNCVLIHNVDQRNSDKDIFGDACDNCLSVLNNDQKDTDGDGRGDACDDDMDGDGIKNILDNCPKFPNRDQRDKDGDGVGDACDSCPDVSNPNQSDVDNDLVGDSCDTNQDSDGDGHQDSTDNCPTVINSAQLDTDKDGIGDECDDDDDNDGIPDLVPPGPDNCRLVPNPAQEDSNSDGVGDICESDFDQDQVIDRIDVCPENAEVTLTDFRAYQTVVLDPEGDAQIDPNWVVLNQGMEIVQTMNSDPGLAVGYTAFNGVDFEGTFHVNTQTDDDYAGFIFGYQDSSSFYVVMWKQTEQTYWQATPFRAVAEPGIQLKAVKSKTGPGEHLRNSLWHTGDTSDQVRLLWKDSRNVGWKDKVSYRWFLQHRPQVGYIRVRFYEGSELVADSGVTIDTTMRGGRLGVFCFSQENIIWSNLKYRCNDTIPEDFQEFQTQNFDRFDNSpecifications
thrombospondin 4 | |
Polyclonal | |
Mouse polyclonal antibody raised against a full-length human THBS4 protein. | |
THBS4 | |
TSP4 | |
Mouse | |
Affinity Purified | |
RUO | |
Primary | |
Human | |
Antibody |
Western Blot | |
Unconjugated | |
PBS with no preservative; pH 7.4 | |
NM_003248.3 | |
THBS4 | |
THBS4 (NP_003239.2, 1 a.a. ∼ 961 a.a) full-length human protein. | |
50 μg | |
Cell Adhesion | |
7060 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.