Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

THEMIS Mouse anti-Human, Alexa Fluor 647, Clone: Q13-1103, BD

Catalog No. BDB563703 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB563703 50 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB563703 Supplier BD Biosciences Supplier No. 563703

Mouse Monoclonal Antibody

The Q13-1103 monoclonal antibody specifically binds to human THEMIS (Thymocyte-expressed molecule involved in selection). The human THEMIS gene encodes a 73-kDa signaling protein that is also known as thymocyte selection pathway associated (TSEPA), signaling phosphoprotein specific for T cells (SPOT) and GRB2-associated protein (GASP). The homologous mouse protein is encoded by the Themis gene. THEMIS mRNA is detected in the thymus, lymph nodes, spleen but not in the bone marrow or nonlymphoid tissues. It is expressed by all thymocytes including double- (high levels) and single-positive αβ T cells, γδ T cells and peripheral T cells (low levels) but not by B cells and NK cells. THEMIS plays a central role in T cell development including positive and negative selection processes in the thymus. It is involved in TCR signaling and gets phosphorylated quickly after TCR stimulation. In the mouse system, -deficient thymocytes showed impaired selection that lead to the production of fewer mature thymocytes. The Q13-1103 antibody crossreacted with mouse Themis in Western blot testing of cellular lysates but did not crossreact when tested by immunofluorescent staining and flow cytometric analysis of cells.

TRUSTED_SUSTAINABILITY

Specifications

Antigen THEMIS
Applications Flow Cytometry
Classification Monoclonal
Clone Q13-1103
Conjugate Alexa Fluor 647
Description TSEPA; THMS1; C6orf190; C6orf207; SPOT; GASP; GRB2-associated protein
Host Species Mouse
Immunogen Human THEMIS Recombinant Protein
Purification Method Affinity Purified
Quantity 50 Tests
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.