Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ anti-Thioredoxin Reductase 1 (TrxR1), Polyclonal,

Catalog No. 07613MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
07-613-MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 07-613-MI Supplier MilliporeSigma™ Supplier No. 07613

Specifications

Antigen Thioredoxin Reductase 1 (TrxR1)
Applications Immunocytochemistry, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation 0.1M Tris-glycine, pH 7.4, 0.15M NaCl, 0.05% sodium azide before the addition of glycerol to 30%
Gene Symbols TXNRD1; TXNR; oxidoreductase; TRXR1; Trxr1; MGC9145; TR; GRIM-12; TR1
Host Species Rabbit
Immunogen peptide corresponding to amino acids 137-150 (C-PHRIKATNNKGKEK) of human thioredoxin reductase 1
Purification Method Affinity Purified
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) NM_003330
Target Species Human, Rat
Content And Storage −20°C for 12 months.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.