Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
mediator complex subunit 16, Mouse, Clone: 2B7, Abnova™
Mouse monoclonal antibody raised against a partial recombinant THRAP5.
Supplier: Abnova Corporation H00010025M02
Description
Sequence: MCDLRRPAAGGMMDLAYVCEWEKWSKSTHCPSVPLACAWSCRNLIAFTMDLRSDDQDLTRMIHILDTEHPWDLHSIPSEHHEAITCLEWDQSGSRLLSADADGQIKCWSMSpecifications
| mediator complex subunit 16 | |
| Monoclonal | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| BC017282 | |
| MED16 | |
| THRAP5 (AAH17282, 1 a.a. ∼ 110 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Signal Transduction | |
| Primary | |
| Human | |
| Antibody |
| ELISA, Immunofluorescence, Western Blot | |
| 2B7 | |
| Mouse monoclonal antibody raised against a partial recombinant THRAP5. | |
| MED16 | |
| DRIP92/THRAP5/TRAP95 | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Yes | |
| 10025 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG2b κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction