Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-Thrombomodulin/BDCA-3, Fluorescein, Clone: 461714, R&D Systems™

Catalog No. FAB3894F
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB3894F 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FAB3894F Supplier R&D Systems Supplier No. FAB3894F
This item is not returnable. View return policy

Applications:

Flow Cytometry

Specifications

Antigen Thrombomodulin/BDCA-3
Applications Flow Cytometry
Classification Monoclonal
Clone 461714
Conjugate Fluorescein
Dilution Flow Cytometry 10 uL/10^6 cells
Formulation Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
Gene Accession No. P15306
Gene Alias BDCA3, BDCA-3, CD141, CD141 antigen, fetomodulin, THBD, THRM, thrombomodulin, TM
Gene Symbols THBD
Host Species Rat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse Thrombomodulin/BDCA-3 Leu17-Ser517 Accession # P15306
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7056
Test Specificity Detects mouse Thrombomodulin/BDCA-3 in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human (rh) Thrombomodulin is observed.
Target Species Mouse
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.