Learn More
MilliporeSigma™ anti-Thrombopoietin Receptor (c-Mpl); Polyclonal; ™ (Upstate™)
Supplier: MilliporeSigma™ 06944/DEL
Description
Moliterno, A. R. and Spivak, J. L.(1999). Posttranslational processing of the thrombopoietin receptor is impaired in polycythemia vera.Blood, 94: 2555-61.
Immunohistochemistry, Immunoprecipitation, Western Blotting
Specifications
Thrombopoietin Receptor/c-Mpl | |
Unconjugated | |
KLH-conjugated, synthetic peptide corresponding to amino acids 615-628 of human TPO (thrombopoietin) receptor (C-ANHSYLPLSYWQQP) | |
RUO | |
Primary | |
Semi-purified |
Polyclonal | |
Rabbit | |
200 μg | |
Cell Signaling | |
Human, Mouse | |