Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thrombospondin-2 Mouse anti-Mouse, Unlabeled, Clone: 4, BD

Catalog No. BDB611150 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB611150 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB611150 Supplier BD Biosciences Supplier No. 611150
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Cell adhesion interactions involve large extracellular glycoproteins such as fibronectin, laminins, and thrombospondins. The thrombospondins (TSPs), a small family of secreted glycoproteins, can be separated into two subfamilies based on structural similarity. TSP-1 and TSP-2 are homotrimers of monomeric 145kDa chains, while TSP-3,4 and 5 are homopentamers of lower MW chains. TSP-1 and TSP-2 interact with several of the same cell surface receptors and have been termed matricellular proteins. However, the functional roles of TSP-1 and TSP-2 are poorly understood. Study of TSP-2 deficient mice has shed some light on the functional role of this protein. Although these mice appear normal, they in fact exhibit a wide variety of irregularities. These include abnormal collagen fiber development which results in fragile skin and highly flexible tendons and ligaments. They also exhibit increased cortical thickness in long bones and abnormal bleeding time. These results, together with the developmental expression pattern of TSP-2, suggest that it functions as a modulator of the cell surface properties of mesenchymal cells which, in turn, affects cell functions such as adhesion and migration.

Host Species: Mouse
Clone: 4
Isotype: IgG1
Species Reactivity: Mouse
Immunogen: Human Thrombospondin-2 aa. 173-295
Formula Weight [Chemical]: 200kDa

Immunofluorescence, Western Blotting

TRUSTED_SUSTAINABILITY

Specifications

Antigen Thrombospondin-2
Applications Western Blot
Classification Monoclonal
Clone 4
Concentration 250μg/mL
Conjugate Unconjugated
Description TSP-2
Formulation Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Human Thrombospondin-2 aa. 173-295
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store undiluted at -20°C.
Isotype IgG1
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.