Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ anti-Thy-1.1 (CD90.1), Clone: OX-7, ™ (Chemicon™)
Supplier: MilliporeSigma™ MAB1406/DEL

Description
Flow Cytometry, Immunocytochemistry, Immunohistochemistry

Specifications
Thy-1.1 (CD90.1) | |
Unconjugated | |
Rat Thy-1 | |
RUO | |
Primary | |
Semi-purified |
Monoclonal | |
Mouse | |
100 μg | |
Inflammation, Immunology | |
Rat | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction