Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thy-1.1 Mouse anti-Rat, Clone: OX-7, MilliporeSigma™

Catalog No. MAB1406MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB1406MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB1406MI Supplier MilliporeSigma Supplier No. MAB1406
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-Thy-1.1 Antibody, clone OX-7 detects level of Thy-1.1 and has been published and validated for use in flow cytometry, immunocytochemistry, immunohistochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Thy-1.1
Applications Flow Cytometry, Immunocytochemistry, Immunohistochemistry
Classification Monoclonal
Clone OX-7
Conjugate Unconjugated
Formulation Protein A Purified mouse immunoglobulin in 20mM sodium phosphate, 250mM NaCl, pH. 7.6, with 0.1% sodium azide as a preservative.
Gene Accession No. NM_006288.2
Gene Symbols THY1, CD90, CDw90, FLJ33325
Host Species Mouse
Immunogen Rat Thy-1
Purification Method Protein A purfied
Quantity 100 μg
Primary or Secondary Primary
Target Species Rat
Content And Storage 2-8°C in undiluted aliquots for up to 12 months. Avoid repeated freeze/thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.