Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thy-1 Mouse anti-Human, Phycoerythrin, Clone: F15-42-1, MilliporeSigma™

Catalog No. CBL415PMI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 Unit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CBL415PMI 1 Unit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CBL415PMI Supplier MilliporeSigma Supplier No. CBL415P
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Detect Thy-1 using this Anti-Thy-1 Antibody, clone F15-42-1, Phycoerythrin conjugated validated for use in flow cytometry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Thy-1
Applications Flow Cytometry
Classification Monoclonal
Clone F15-42-1
Conjugate PE
Formulation The conjugate is supplied as a 100 test vial in phosphate buffered saline containing 10mM sodium azide and 1mg/ml bovine serum albumin. For flow cytometry we recommend using 10μL of conjugate per test.
Gene Accession No. NM_006288.2
Gene Symbols THY1, CD90, CDw90, FLJ33325
Host Species Mouse
Quantity 1 Unit
Primary or Secondary Primary
Target Species Human
Content And Storage 2-8°C, protect from prolonged exposure to light. Do not Freeze.
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.