Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thymidylate Kinase/TMPK Rabbit anti-Human, Polyclonal, MilliporeSigma™

Catalog No. ABC458MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ABC458MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ABC458MI Supplier MilliporeSigma Supplier No. ABC458

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Rabbit Polyclonal Antibody

Anti-Thymidylate Kinase/TMPK Antibody is a rabbit polyclonal antibody, validated for use in western blotting and immunoprecipitation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Thymidylate Kinase/TMPK
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Purified rabbit polyclonal in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Symbols DTYMK; CDC8; TMPK; TYMK
Host Species Rabbit
Immunogen GST-tagged recombinant protein corresponding to human Thymidylate Kinase/TMPK.
Purification Method Protein A purified
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) NP_036277
Target Species Human
Content And Storage 2-8°C for 12 months
Form Purified
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.