Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thymidylate Synthase Mouse anti-Human, Mouse, Rat, Clone: TS106, MilliporeSigma™

Catalog No. MAB4130MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB4130MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB4130MI Supplier MilliporeSigma Supplier No. MAB4130
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Use Anti-Thymidylate Synthase Antibody, clone TS106 (Mouse Monoclonal Antibody) validated in ELISA, flow cytometry, immunofluoresence, immunohistochemistry, immunohistochemistry (paraffin), immunoprecipitation, western blotting to detect Thymidylate Synthase also known as Thymidylate synthase.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Thymidylate Synthase
Applications ELISA, Flow Cytometry, Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone TS106
Conjugate Unconjugated
Formulation Purified mouse monoclonal IgG1 in 0.02M PBS with 0.25M NaCl, pH=7.6, 0.1% sodium azide.
Gene Accession No. NM_001071.1
Gene Symbols TYMS, TS, Tsase, TSase, TMS, HsT422, MGC88736, EC 2.1.1.45
Host Species Mouse
Immunogen Recombinant Thymidylate Synthase.
Purification Method Protein A purfied
Quantity 100 μg
Primary or Secondary Primary
Target Species Human, Mouse, Rat
Content And Storage 2-8°C for one year
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.