Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TIAR Mouse, Unlabeled, Clone: 6, BD

Catalog No. BDB610352 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB610352 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB610352 Supplier BD Biosciences Supplier No. 610352
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

TIAR is an RNA-binding protein related to TIA-1. Both proteins are candidate effectors of apoptotic cell death and consist of three N-terminal RNA-recognition motifs (RRM) and a C-terminal protein-interaction domain (PID). Unlike TIA-1, which is localized in the granules of cytotoxic lymphocytes, TIAR is predominantly found in the nucleus of many different cell types. Two related isoforms (42 kDa and 50 kDa) of TIAR have been identified. It has been demonstrated that TIAR can trigger DNA fragmentation in permeabilized thymocytes, suggesting that it may be an effector of apoptotic cell death. During Fas-mediated apoptosis, TIAR shows a rapid translocation from the nucleus tothe cytoplasm. This redistribution of TIAR appears to be a specific event and possibly a general feature of the apoptotic program.

TRUSTED_SUSTAINABILITY

Specifications

Antigen TIAR
Applications Western Blot
Classification Monoclonal
Clone 6
Concentration 250μg/mL
Conjugate Unconjugated
Formulation Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Canine, Frog, Human, Mouse, Rat
Content And Storage Store undiluted at -20°C.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.