Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tie-2 Rabbit anti-Human, Mouse, Rat, Polyclonal, MilliporeSigma™

Catalog No. 071431MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
07-143-1MI 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 07-143-1MI Supplier MilliporeSigma Supplier No. 071431
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This Anti-Tie-2 Antibody, N-terminus is validated for use in western blotting for the detection of Tie-2.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Tie-2
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Affinity purified immunoglobulin. Liquid in 100mM Tris, pH 7.5, containing 0.2% BSA and 0.05% sodium azide.
Gene Symbols CD202B; CD202b; EC 2.7.10.1 ; OTTHUMP00000021167; TIE-2 ; TIE2; VMCM; VMCM1; hTIE2
Host Species Rabbit
Immunogen 18 amino acid peptide sequence within the extracellular, N-terminus of human Tie-2
Purification Method Affinity Purified
Quantity 50 μg
Primary or Secondary Primary
Gene ID (Entrez) NP_000450
Target Species Human, Mouse, Rat
Content And Storage −20°C in undiluted aliquots for 12 months. Avoid repeated freeze/thaw cycles.
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.