Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ anti-Tie2 Clone: Ab33,

Catalog No. 61020550UG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
50 μg
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
61-020-550UG 50 μg Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 61-020-550UG Supplier MilliporeSigma™ Supplier No. 61020550UG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Specifically detects Tie2 Clone: Ab33 in Human, Mouse, Porcine, Rat samples, and it is validated for Immunohistochemistry (Frozen), Immunoprecipitation, Immuno Blotting

Angiopoietin-1 and its naturally occurring inhibitor, angiopoietin-2, exert their effects by inducing autophosphorylation of their signaling receptor Tie2. Phosphorylation of the receptor occurs during vasculogenesis and is required for its activity in regulating endothelial cell proliferation, differentiation and guiding the proper patterning of endothelial cells during blood vessel formation. Tie-2 is a transmembrane receptor-tyrosine kinase that is expressed by endothelial cells and primitive hematopoietic tissues. Tie2/Tek has been shown to be critical for endothelial cell survival, which is evidenced in part by the constitutive phosphorylation that is observed in quiescent endothelium. More recently a multifunction docking site has been identified on the receptor (Tyr1100) that serves as a binding site for other partners including Grb2, Grb7, and p85 and acting via an SH2 domain interaction.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Tie2
Applications Immunoblot, Immunohistochemistry (Frozen), Immunoprecipitation
Classification Monoclonal
Clone Ab33
Conjugate Unconjugated
Formulation In 150mM NaCl, 70mM Tris-glycine, 30% glycerol, pH 7.4.
Host Species Mouse
Immunogen a recombinant protein consisting of amino acids 1-745 of human Tie2 fused to a His·Tag™ sequence
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Recognizes the ∽140-45kDa Tie2 protein.
Target Species Human, Mouse, Pig, Rat
Content And Storage −20°C
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.