Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TIE2 (pY1102) Mouse, Alexa Fluor 488, Clone: K93-754, BD

Catalog No. BDB560053 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB560053 50 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB560053 Supplier BD Biosciences Supplier No. 560053
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

TIE2 (Tyrosine kinase with Immunoglobulin-like and EGF-like domains unical Endothelial Kinase), is an endothelial and hematopoietic cell-specific receptor tyrosine kinase (RTK) that is critical to the development and maintenance of the vasculature and highly conserved among vertebrate species. The angiopoietins are ligands of TIE2, and the abnormal vascular growth that often occurs in solid tumors is a result of disruptions in the coordinated actions of the angiopoietins, TIE2, and the closely related TIE1 RTK. Upon activation by angiopoietins, TIE2 autophosphorylates at least 2 tyrosines in its protein kinase domain and at least 3 tyrosines in its C-terminal tail. Phosphorylation at tyrosine 1102 (Y1102) leads to activation of the Akt signal transduction pathway, enhancing endothelial cell survival. The phosphorylated Y1102 also recruits and activates the src homology 2 domain-containing adaptor proteins Grb2 and ShcA, affecting endothelial cell adhesion and motility.

The K93-754 monoclonal antibody recognizes the phosphorylated Y1102 in the C-terminal tail of activated TIE2. The orthologous phosphorylation site in mouse TIE2 is Y1100.

TRUSTED_SUSTAINABILITY

Specifications

Antigen TIE2 (pY1102)
Applications Flow Cytometry
Classification Monoclonal
Clone K93-754
Conjugate Alexa Fluor 488
Description TEK, VMCM, CD202b
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Phosphorylated Human TIE2 Peptide
Purification Method Affinity Purified
Quantity 50 Tests
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human, Mouse
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG2a κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.