Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tie2/TEK Mouse anti-Human, Mouse, Rat, Pig, Clone: Ab33, MilliporeSigma™

Catalog No. 05584MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
05-584-MI 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 05-584-MI Supplier MilliporeSigma Supplier No. 05584
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This Anti-Tie2/TEK Antibody, clone Ab33 is validated for use in western blot, immunoprecipitation, immunohistochemistry for the detection of Tie2/TEK.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Tie2/TEK
Applications Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone Ab33
Conjugate Unconjugated
Formulation Purified mouse monoclonal IgG1κ in buffer containing 0.1MTris-glycine, pH 7.4, 0.15MNaCl, 0.05% sodium azide before the addition of glycerol to 30%. Liquid at −20°C.
Gene Accession No. NM_000459
Gene Symbols TEK, CD202B, TIE2, VMCM1, VMCM, hTIE2, CD202b, TIE-2
Host Species Mouse
Immunogen Fusion protein corresponding to residues 1-745 of human Tie2/TEK with a C-terminal His6-tag purified from infected SF-9 cells. Clone Ab33
Purification Method Protein G purfied
Quantity 200 μg
Primary or Secondary Primary
Target Species Human, Mouse, Pig, Rat
Content And Storage Stable for 1 year at from date of receipt.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.