Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TIGIT Mouse anti-Mouse, PE, Clone: 1G9, BD

Catalog No. BDB565168 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB565168 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB565168 Supplier BD Biosciences Supplier No. 565168
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The 1G9 monoclonal antibody specifically binds to T cell immunoreceptor with Ig and ITIM domains (TIGIT) which is also known as, V-set and transmembrane domain-containing protein 3 (Vstm3). TIGIT is a 26 kDa, type I transmembrane glycoprotein that belongs to the poliovirus receptor (PVR) family. TIGIT is expressed on activated and memory CD4+ T cells and CD8+ T cells, follicular T helper cells, regulatory T cells, and NK cells. Its ligands include the poliovirus receptor (PVR, aka, CD155), poliovirus receptor-related protein 2 (PVRL2, aka, CD112), and poliovirus receptor-related protein 3 (PVRL3, CD113). TIGIT functions as a negative regulator of T cell and NK cell responses including proliferation, cytotoxicity, and the production of proinflammatory cytokines. TIGIT-deficient mice are reportedly more susceptible to developing autoimmune diseases in certain model systems.

TRUSTED_SUSTAINABILITY

Specifications

Antigen TIGIT
Applications Flow Cytometry
Classification Monoclonal
Clone 1G9
Concentration 0.2mg/mL
Conjugate PE
Description Tigit; Vstm3; T cell immunoreceptor with Ig and ITIM domains
Formulation Aqueous buffered solution containing ≤0.09% sodium azide
Host Species Mouse
Immunogen Mouse TIGIT Recombinant Protein
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.