Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TIM-3 Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

$181.90 - $426.00

Specifications

Antigen TIM-3
Concentration 0.5 mg/mL
Dilution Western Blot 1-3 ug/ml
Applications Western Blot
Classification Polyclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB10056422T
SDP
View Documents
Novus Biologicals
NB10056422SS
0.025 mg
Each for $181.90
Only null left
Add to Cart
 
NB10056422
SDP
View Documents
Novus Biologicals
NB10056422
0.1 mg
Each for $426.00
Only null left
Add to Cart
 
Description

Description

TIM-3 Polyclonal specifically detects TIM-3 in Mouse, Rat samples. It is validated for Western Blot.
Specifications

Specifications

TIM-3
Western Blot 1-3 ug/ml
Polyclonal
Purified
RUO
Mouse, Rat
CD366, FLJ14428, HAVcr-2, hepatitis A virus cellular receptor 2, kidney injury molecule-3, T cell immunoglobulin mucin 3, T cell immunoglobulin mucin-3, T-cell immunoglobulin and mucin domain-containing protein 3, TIM 3, Tim-3, TIM3 T-cell membrane protein 3, TIMD-3, TIMD3KIM-3
HAVCR2
IgG
Protein G purified
The NCBI accession number for TIM-3 is AAL35776.
0.5 mg/mL
Western Blot
Unconjugated
Rabbit
Cancer, Immunology, Innate Immunity, Neuroscience
PBS containing 0.05% BSA with 0.05% Sodium Azide
84868
A portion of amino acids 100-200 of mouse TIM-3 protein were used as the immunogen.
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.