Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TIM-4 Rat anti-Mouse, PE, Clone: RMT4-54, BD

Catalog No. BDB564147 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB564147 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB564147 Supplier BD Biosciences Supplier No. 564147
Add to Cart
Add to Cart

Rat Monoclonal Antibody

The RMT4-54 monoclonal antibody specifically binds to TIM-4. TIM-4 is encoded by Timd4 (T cell immunoglobulin and mucin domain containing 4). TIM-4 is also known as Spleen, mucin-containing, knockout of lymphotoxin protein (SMUCKLER). TIM-4 is a single-pass type I membrane transmembrane glycoprotein belonging to the TIM family of the immunoglobulin superfamily. TIM-4 is expressed by macrophages and at low levels by dendritic cells. TIM-4 is a phosphatidylserine receptor that enhances the phagocytosis of apoptotic cells. It can serve as a receptor for TIM-1, also known as the Hepatitis A virus cellular receptor 1 (Havcr1).

TRUSTED_SUSTAINABILITY

Specifications

Antigen TIM-4
Applications Flow Cytometry
Classification Monoclonal
Clone RMT4-54
Concentration 0.2mg/mL
Conjugate PE
Description TIMD-4, SMUCKLER, Tim4, Timd4
Host Species Rat
Immunogen Mouse Tim-4 Recombinant Protein
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG2a κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.