Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tim23 Mouse, Unlabeled, Clone: 32, BD

Catalog No. BDB611223 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
150 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB611223 150 μg
BDB611222 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB611223 Supplier BD Biosciences Supplier No. 611223
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Mitochondria, the site of cellular energy production, must import all proteins necessary for their function. Import is mediated by two mechanisms: the translocase of the outer membrane (Tom) and the translocase of the inner membrane (Tim). Tim23 and Tim17 are integral membrane proteins that associate to form the import channel for mitochondrial preproteins that contain N-terminal hydrophilic sequences. They also associate with Tim44, an adaptor for the membrane binding of mtHsp70, a matrix heat shock protein, which drives the import of the
processed preprotein. The N-terminal intermembrane space domain of Tim23 contains a leucine zipper motif and mediates the formation of a Tim23 dimer. As an imported protein passes through the TOM machinery, its N-terminal matrix targeting sequence interacts with the Tim23 dimer. This induces the dissociation of the dimer and initiation of inner membrane translocation of the presequence. In addition to its 9kDa N-terminal hydrophilic segment, Tim23 contains a 14kDa hydrophobic domain with four predicted membrane spans. Thus, Tim23 is an important integral membrane component of the mitochondrial protein translocation machinery.

Host Species: Mouse
Clone: 32
Isotype: IgG2a
Species Reactivity: Mouse
Immunogen: Rat Tim23 aa. 5-126
Formula Weight [Chemical]: 23kDa

Immunofluorescence, Western Blotting

TRUSTED_SUSTAINABILITY

Specifications

Antigen Tim23
Applications Western Blot
Classification Monoclonal
Clone 32
Concentration 250μg/mL
Conjugate Unconjugated
Formulation Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Rat Tim23 aa. 5-126
Purification Method Affinity Purified
Quantity 150 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Human, Mouse, Rat
Content And Storage Store undiluted at -20°C.
Isotype IgG2a
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.