Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tim3 Mouse anti-Mouse, Clone: 5D12, MilliporeSigma™

Catalog No. MABF73MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABF73MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABF73MI Supplier MilliporeSigma Supplier No. MABF73
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-Tim3 Antibody, clone 5D12 is an antibody against Tim3 for use in flow cytometry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Tim3
Applications Flow Cytometry
Classification Monoclonal
Clone 5D12
Conjugate Unconjugated
Formulation Purified mouse monoclonal IgG1κ in buffer containing 0.1MTris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Accession No. NP_599011
Gene Symbols Havcr2, Tim3, Timd3
Host Species Mouse
Immunogen Recombinant protein corresponding to full length mouse Tim3.
Purification Method Protein G Purified
Quantity 100 μL
Primary or Secondary Primary
Target Species Mouse
Content And Storage 2-8°C for one year
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.