Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TIMM13, Rabbit, Monoclonal Antibody, Abnova™

Catalog No. 89210822 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 mg x up to 3 Abs
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-210-822 1 mg x up to 3 Abs
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-210-822 Supplier Abnova Corporation Supplier No. H00026517K

Includes

Up to three rabbit IgG clones of 1mg each will be delivered to customer.

Rabbit monoclonal antibody raised against a human TIMM13 peptide using ARM Technology.

This gene encodes a translocase with similarity to yeast mitochondrial proteins that are involved in the import of metabolite transporters from the cytoplasm and into the mitochondrial inner membrane. The encoded protein and the TIMM8a protein form a 70kDa complex in the intermembrane space. This gene is in a head-to-tail orientation with the gene for lamin B2. [provided by RefSeq

Library Construction: Nonfusion antibody library from rabbit spleen (ARM Technology)
Expression: Overexpression vector and transfection into 293H cell line

Order Info

  1. Customer may provide cell or tissue lysate for antibody screening.
  2. Rabbit monoclonal antibody generated by ARM technology is amenable to antibody engineering including F(ab)2, IgG, scFv and different Fc and non-Fc conjugates per customer request.

Specifications

Antigen TIMM13
Applications ELISA, Western Blot
Classification Monoclonal
Concentration 1mg
Conjugate Unconjugated
Description translocase of inner mitochondrial membrane 13 homolog (yeast)
Formulation In 1X PBS, pH 7.4
Gene TIMM13
Gene Accession No. TIMM13
Gene Alias TIM13, TIM13B, TIMM13A, TIMM13B, ppv1
Gene Symbols TIMM13
Host Species Rabbit
Immunogen A synthetic peptide of human TIMM13
Quantity 1 mg x up to 3 Abs
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26517
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.