Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TIMM8A, Mouse, Clone: 1A12, Abnova™

Catalog No. 89123149 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-123-149 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-123-149 Supplier Abnova Corporation Supplier No. H00001678M04
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a full-length recombinant TIMM8A.

This translocase is involved in the import and insertion of hydrophobic membrane proteins from the cytoplasm into the mitochondrial inner membrane. The gene is mutated in Mohr-Tranebjaerg syndrome/Deafness Dystonia Syndrome (MTS/DDS) and it is postulated that MTS/DDS is a mitochondrial disease caused by a defective mitochondrial protein import system. Defects in this gene also cause Jensen syndrome; an X-linked disease with opticoacoustic nerve atrophy and muscle weakness. This protein, along with TIMM13, forms a 70kDa heterohexamer. Alternative splicing results in multiple transcript variants encoding distinct isoforms

Sequence: MLLNDKWVNEEIKKKIEKCLETNDNGNTTYQNLWDTAKAVVRGKFIAISTYIKKEEKLQINNLTMNLIELEN

Specifications

Antigen TIMM8A
Applications ELISA, Western Blot
Classification Monoclonal
Clone 1A12
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a full-length recombinant TIMM8A.
Formulation PBS with no preservative; pH 7.4
Gene TIMM8A
Gene Accession No. BC005236
Gene Alias DDP/DDP1/DFN1/MGC12262/MTS
Gene Symbols TIMM8A
Host Species Mouse
Immunogen TIMM8A (AAH05236, 1 a.a. ∼ 72 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1678
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Liquid
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.