Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TIMP-1 Rabbit anti-Bovine, Horse, Ferret, Rat, Rabbit, Sheep, Polyclonal, MilliporeSigma™

Catalog No. AB770MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AB770MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AB770MI Supplier MilliporeSigma Supplier No. AB770
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Anti-TIMP-1 Antibody detects level of TIMP-1 and has been published and validated for use in ELISA, western blot, immunohistochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TIMP-1
Applications ELISA, Immunohistochemistry, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Purified immunoglobulin. Liquid in PBS containing 0.02% azide.
Gene Symbols TIMP1; TIMP; FLJ90373; CLGI; EPA; EPO; TIMP-1; HCI
Host Species Rabbit
Immunogen Full length purified human fibroblast tissue inhibitor of metalloproteinases {Clark, IM (1991) Matrix 11:76-85}.
Purification Method Affinity Purified
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) NM_003254.2
Target Species Bovine, Equine, Ferret, Rabbit, Rat, Sheep
Content And Storage 2-8°C in undiluted aliquotes for 3 months, −20°C in undiluted aliquots for up to 6 months, Avoid Freeze/Thaw Cycles
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.