Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TIMP-2 Mouse anti-Bovine, Guinea Pig, Goat, Human, Mouse, Rat, Rabbit, Clone: 67-4H11, MilliporeSigma™

Catalog No. MAB3310MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB3310MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB3310MI Supplier MilliporeSigma Supplier No. MAB3310
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-TIMP-2 Antibody, clone 67-4H11 detects level of TIMP-2 and has been published and validated for use in ELISA, western blot, immunohistochemistry (paraffin).
TRUSTED_SUSTAINABILITY

Specifications

Antigen TIMP-2
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 67-4H11
Conjugate Unconjugated
Formulation Liquid in 0.1M Na-Phosphate buffer, pH 7.0 containing 2% protease free bovine serum albumin.
Gene Accession No. NM_003255.4
Gene Symbols TIMP2, TIMP-2, CSC-21K
Host Species Mouse
Immunogen Synthetic peptide corresponding to amino acids 178-193 of human TIMP-2 (numbered from the propeptide) YRGAAPPKQEFLDIED
Quantity 100 μg
Primary or Secondary Primary
Target Species Bovine, Goat, Guinea Pig, Human, Mouse, Rabbit, Rat
Content And Storage −20°C in undiluted aliquots for up to 12 months.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.