Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TIMP-2 (CT) Rabbit anti-Human, Mouse, Rat, Bovine, Pig, Horse, Canine, Rabbit, Polyclonal, MilliporeSigma™

Catalog No. AB2965MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AB2965MI 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AB2965MI Supplier MilliporeSigma Supplier No. AB2965

Rabbit Polyclonal Antibody

Detect TIMP-2 (C-terminus) using this Anti-TIMP-2 (C-terminus) Antibody validated for use in western blot, immunohistochemistry (paraffin).
TRUSTED_SUSTAINABILITY

Specifications

Antigen TIMP-2 (CT)
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Purified rabbit polyclonal in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Symbols TIMP2; TIMP-2; CSC-21K
Host Species Rabbit
Immunogen KLH-conjugated linear peptide corresponding to the C-terminus of human TIMP-2.
Purification Method Affinity Purified
Quantity 50 μg
Primary or Secondary Primary
Gene ID (Entrez) NP_003246
Target Species Bovine, Canine, Equine, Human, Mouse, Pig, Rabbit, Rat
Content And Storage 2-8°C for 12 months
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.