Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TIMP-3 Mouse anti-Human, Rabbit, Clone: 136-13H4, MilliporeSigma™

Catalog No. MAB3318MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB3318MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB3318MI Supplier MilliporeSigma Supplier No. MAB3318
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-TIMP-3 Antibody, a.a. 170-188, clone 136-13H4 is an antibody against TIMP-3 for use in immunohistochemistry (paraffin) and western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TIMP-3
Applications Immunohistochemistry, Western Blot
Classification Monoclonal
Clone 136-13H4
Conjugate Unconjugated
Formulation Liquid in 0.1Msodium phosphate buffer, pH 7.0 containing 2% protease free bovine Serum albumin.
Gene Accession No. NM_000362.4
Gene Symbols TIMP3, HSMRK222, K222, K222TA2, TIMP-3, SFD
Host Species Mouse
Immunogen Oligopeptide of SWYRGWAPPDKSIINATDP (residue 170-188) on human tissue inhibitor of metalloproteinases-3.
Purification Method Protein A purfied
Quantity 100 μg
Primary or Secondary Primary
Target Species Human, Rabbit
Content And Storage −20°C in undiluted aliquots for up to 12 months. Avoid repeated freeze/thaw cycles.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.