Learn More
anti-TIMP-3, PE, Clone: 277128, R&D Systems™
Description
Applications:
Flow Cytometry (Intracellular Staining)
Specifications
Specifications
| Antigen | TIMP-3 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | 277128 |
| Conjugate | R-PE |
| Dilution | Intracellular Staining by Flow Cytometry 10 uL/10^6 cells |
| Formulation | Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide |
| Gene Accession No. | P35625 |
| Gene Alias | HSMRK222, K222, K222TA2, metalloproteinase inhibitor 3, MIG-5 protein, Protein MIG-5, pseudoinflammatory), SFD, TIMP metallopeptidase inhibitor 3, TIMP3, TIMP-3, tissue inhibitor of metalloproteinase 3 (Sorsby fundus dystrophy, Tissue inhibitor of metalloproteinases 3 |
| Gene Symbols | TIMP3 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.