Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

tubulointerstitial nephritis antigen, Mouse, Purified MaxPab™ Polyclonal Antibody, Abnova™

Catalog No. 89017058 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-017-058 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-017-058 Supplier Abnova Corporation Supplier No. H00027283B01P
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a full-length human TINAG protein.

TINAG is a basement membrane glycoprotein initially identified as a target of antibodies in some forms of immunologically mediated tubulointerstitial nephritis.[supplied by OMIM

Sequence: MWTGYKILIFSYLTTEIWMEKRYLSQREVDLEAYFTRNHTVLQGTRFKRAIFQGQYCRNFGCCEDRDDGCVTEFYAANALCYCDKFCDRENSDCCPDYKSFCREEKEWPPHTQPWYPEGCFKDGQHYEEGSVIKENCNSCTCSGQQWKCSQHVCLVRSELIEQVNKGDYGWTAQNYSQFWGMTLEDGFKFRLGTLPPSLMLLSMNEMTASLPATTDLPEFFVASYKWPGWTHGPLDQKNCAASWAFSTASVAADRIAIQSKGRYTANLSPQNLISCCAKNRHGCNSGSIDRAWWYLRKRGLVSHACYPLFKDQNATNNGCAMASRSDGRGKRHATKPCPNNVEKSNRIYQCSPPYRVSSNETEIMKEIMQNGPVQAIMQVHEDFFHYKTGIYRHVTSTNKESEKYRKLQTHAVKLTGWGTLRGAQGQKEKFWIAANSWGKSWGENGYFRILRGVNESDIEKLIIAAWGQLTSSDEP

Specifications

Antigen tubulointerstitial nephritis antigen
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a full-length human TINAG protein.
Formulation PBS with no preservative; pH 7.4
Gene TINAG
Gene Accession No. NM_014464.2
Gene Alias TIN-AG/TIN1/TIN2
Gene Symbols TINAG
Host Species Mouse
Immunogen TINAG (NP_055279.2, 1 a.a. ∼ 476 a.a) full-length human protein.
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Cell Adhesion
Primary or Secondary Primary
Gene ID (Entrez) 27283
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.