Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TIP49b Mouse, Unlabeled, Clone: 42, BD

Catalog No. BDB612482 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB612482 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB612482 Supplier BD Biosciences Supplier No. 612482
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Processes involving DNA transcription, replication and repair are necessary in maintaining normal cell function. For each of these processes, the double stranded NA must first be unwound. This is achieved in an ATP-dependent manner by enzymes termed DNA helicases. Reptin52/TIP49b is a DNA helicase that directly interacts with Pontin52/TIP49a. Reptin52/TIP49b and Pontin52/TIP49a belong to a family of highly conserved proteins that share homology with the bacterial protein RuvB, a helicase involved in double-strand break repair and homologous recombination. Both are TATA binding protein (TBP) interacting proteins that bind to β-Catenin and are among the 12 polypeptides that comprise the INO80 chromatin-remodeling complex. It has also been demonstrated that Reptin52/TIP49b and Pontin52/TIP49a can be found in a complex with the proto- ncogene c-myc. Reptin52/TIP49b and Pontin52/TIP49a have antagonistic roles as Reptin/TIP49b represses gene activation mediated by β-Catenin and c-myc, whereas Pontin52/TIP49a stimulates gene activation. Thus, Reptin52/TIP49b and Pontin52/TIP49a are transcriptional co-factors that work together to help control multiple pathways.

Immunofluorescence, Western Blotting

TRUSTED_SUSTAINABILITY

Specifications

Antigen TIP49b
Applications Western Blot
Classification Monoclonal
Clone 42
Concentration 250μg/mL
Conjugate Unconjugated
Formulation Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Human Reptin 52/TIF-496 aa. 180-228
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Canine, Chicken, Human, Mouse, Rat
Content And Storage Store undiluted at -20°C.
Isotype IgG1
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.