Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ anti-TIP60, Polyclonal

Catalog No. DR104150UL Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
50 μL
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
DR104150UL 50 μL Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. DR104150UL Supplier MilliporeSigma™ Supplier No. DR104150UL

Rabbit Polyclonal Antibody

Specifically detects TIP60 Clone: in Human samples, and it is validated for Immunoprecipitation

TIP60 (accession no. Q92993) is a 60kDa protein that is a member of the MYST family, which is a distinct class of histone acetyl-transferases (HAT). TIP60 was initially identified as a protein that interacts with the HIV protein, Tat, and shares no homology with the HATs, CBP/p300, or PCAF. TIP60 was recently shown to modulate DNA repair and p53 function. TIP60 also directly acetylatesγ-H2AX and, together with the cofactor, TRRAP, binds to the chromatin surrounding sites of DNA double-stranded breaksin vivo. Furthermore, TIP60 associates with MYC and recruits it to the chromatin, which results in MYC-induced actetylation of histone H4. TIP60 directly associates with p53 and specifically acetylates it at K120, modulating the decision between cell cycle arrest and apoptosis. TIP60 acts as a transcription coactivator of p53 that enhances the activation of p21 and puma, leading to cell growth arrest and apoptosis. K120 actetylation is essential for p53-mediated puma activation and apoptosis, but is dispensable for both p53-mediated growth arrest, through p21 activation, and p53/MDM2 interaction. K120 is not acetylated by p300 and its actetylation by TIP60 does not affect the stability or DNA binding activity of p53.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TIP60
Applications Immunoprecipitation
Classification Polyclonal
Conjugate Unconjugated
Formulation Undiluted serum.
Host Species Rabbit
Immunogen recombinant, human TIP60 protein
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Recognizes the ∽60kDa TIP60 protein in Saos-2 cells.
Target Species Human
Content And Storage −20°C
Form Undiluted
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.