Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ anti-Tissue Factor Clone: TF9-10H10,

Catalog No. 612161500UG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
500 μg
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
61-216-1500UG 500 μg Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 61-216-1500UG Supplier MilliporeSigma™ Supplier No. 612161500UG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Specifically detects Tissue Factor Clone: TF9-10H10 in Human, Primate samples, and it is validated for Dot Blot, ELISA, Flow Cytometry, Immunofluorescence, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immuno Blotting

Tissue factor (TF) is the membrane receptor for coagulation factors VII and VIIa and is a cell-surface initiator of coagulation. It is the major molecule of this type and is critical for controlling hemostasis, thrombosis, and inflammation. TF is not generally present in or on cells within the vasculature, but following vascular injury, blood can come into contact with TF expressed by extravascular cells, resulting in activation of the coagulation cascade.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Tissue Factor
Applications Dot Blot, ELISA, Flow Cytometry, Immunoblot, Immunoblot, Immunofluorescence, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone TF9-10H10
Conjugate Unconjugated
Formulation Lyophilized from 200mM mannitol, 150mM PBS, pH 7.4.
Host Species Mouse
Immunogen purified apoprotein of human tissue factor
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Recognizes the tissue factor protein.
Target Species Human, Primate
Content And Storage −20°C
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.