Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ anti-Tissue Plasminogen Activator Clone: GMA-043,

Catalog No. GF53100UG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
GF53100UG 100 μg Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. GF53100UG Supplier MilliporeSigma™ Supplier No. GF53100UG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Specifically detects Tissue Plasminogen Activator Clone: GMA-043 in Human samples, and it is validated for ELISA, Immuno Blotting

Epidemiological studies have demonstrated that levels of plasma fibrinogen, von Willebrand factor (vWf), plasminogen activator inhibitor-1 (PAI-1) and tissue-type plasminogen activator (tPA) are associated with the incidence of vascular disease. Von Willebrand factor (vWF) is a large, multimeric glycoprotein mediating platelet adhesion at sites of endothelial damage. The process of acute release of tissue-type plasminogen activator (tPA) is important in locally speeding up fibrinolysis. Acutely released tPA and constitutively secreted tPA were liberated from different pathways in human umbilical vein endothelial cells. Plasma levels of tissue plasminogen activator to plasminogen activator/ inhibitor-1 complex, in addition to von Willebrand factor, are significant risk markers for recurrent myocardial infarction.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Tissue Plasminogen Activator
Applications ELISA, Immunoblot, Immunoblot
Classification Monoclonal
Clone GMA-043
Conjugate Unconjugated
Formulation In 150mM NaCl, 10mM sodium phosphate, 1% mannitol, pH 7.4.
Host Species Mouse
Immunogen purified human tissue plasminogen activator (tPA)
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Recognizes the ∽60kDa tPA protein.
Target Species Human
Content And Storage −20°C
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.