Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
tPA, clone: T-1, Invitrogen™
Mouse Monoclonal Antibody
Supplier: Thermo Scientific MONT102
Description
Tissue plasminogen activator (tPA) is a serine protease which converts the abundant proenzyme plasminogen to active plasmin. It plays a key role in thrombolysis and several neurobiological processes.
MON T-1-02 is specific for human tPA. No reaction is seen with human urokinase plasminogen activator (uPA) when tested by ELISA, immunoblotting and enzyme inhibition and no reaction with any other human plasma proteins is seen when tested by immunoblotting.MON T-1-02 has successfully been used in ELISA, Western blot (non-reducing conditions) and immunohistochemistry (frozen) applications.The immunogen used to produce MON T-1-02 is native human tPA.NOTE: Concentration is lot-dependent and can vary from 0.85-1.15 mg/ml
Specifications
tPA | |
Monoclonal | |
0.95mg/mL | |
PLAT | |
DKFZp686I03148, T-PA, TPA | |
Mouse | |
Protein A | |
RUO | |
5327 | |
4° C, store in dark, DO NOT FREEZE! | |
Liquid |
ELISA, Immunohistochemistry (Frozen), Western Blot | |
T-1 | |
Unconjugated | |
P00750 | |
PLAT | |
Native human tPA. | |
200 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction