Learn More
Description
Specifications
Specifications
| Antigen | TK |
| Applications | Immunoprecipitation |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Host Species | Mouse |
| Immunogen | A proprietary formulation containing tyrosine kinase (TK) antibody at 20X concentration for use in KinEASE™ fluorescence polarization assays |
| Quantity | 237 μL |
| Primary or Secondary | Primary |
| Target Species | Mouse |
| Content And Storage | −20°C for up to 6 months. |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.