Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

thymidine kinase 1, soluble, Mouse, Clone: 3F9, Abnova™

Catalog No. 89001022 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-001-022 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-001-022 Supplier Abnova Corporation Supplier No. H00007083M05
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a full-length recombinant TK1.

Sequence: MSCINLPTVLPGSPSKTRGQIQVILGPMFSGKSTELMRRVRRFQIAQYKCLVIKYAKDTRYSSSFCTHDRNTMEALPACLLRDVAQEALGVAVIGIDEGQFFPDIVEFCEAMANAGKTVIVAALDGTFQRKPFGAILNLVPLAESVVKLTAVCMECFREAAYTKRLGTEKEVEVIGGADKYHSVCRLCYFKKASGQPAGPDNKENCPVPGKPGEAVAARKLFAPQQILQCSPAN

Specifications

Antigen thymidine kinase 1, soluble
Applications ELISA, Western Blot
Classification Monoclonal
Clone 3F9
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a full length recombinant TK1.
Formulation PBS with no preservative; pH 7.4
Gene TK1
Gene Accession No. BC007986
Gene Alias TK2
Gene Symbols TK1
Host Species Mouse
Immunogen TK1 (AAH07986.1, 1 a.a. ∼ 234 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Stem Cell Biology
Whole Molecule Yes
Primary or Secondary Primary
Gene ID (Entrez) 7083
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.