Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Tks4 Rabbit anti-Human, Mouse, Polyclonal, MilliporeSigma™

Catalog No. 09267MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
09-267-MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 09-267-MI Supplier MilliporeSigma Supplier No. 09267

Rabbit Polyclonal Antibody

This Anti-human Tks4 Antibody is validated for use in western blot, immunoprecipitation for the detection of human Tks4.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Tks4
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Purified in 0.1M Tris-Glycine, pH 7.4, 150mM NaCl with 0.05% NaN3.
Gene Accession No. A1X283
Host Species Rabbit
Immunogen Recombinant protein containing amino acids 431-595 of human Tks4.
Purification Method Protein A purfied
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) FAD49, FLJ20831, HOFI, KIAA1295
Target Species Human, Mouse
Content And Storage 2-8°C for up to 12 months
Form Purified
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.