Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TKS5 (SH3 #4) Rabbit anti-Human, Mouse, Polyclonal, MilliporeSigma™

Catalog No. 09268MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
09-268-MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 09-268-MI Supplier MilliporeSigma Supplier No. 09268
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Anti-TKS5 (SH3 #4) Antibody is an antibody against TKS5 (SH3 #4) for use in immunocytochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TKS5 (SH3 #4)
Applications Immunocytochemistry
Classification Polyclonal
Conjugate Unconjugated
Formulation Purified in 0.1M Tris-Glycine (pH7.4) 150mM NaCl with 0.05% NaN3.
Gene Symbols FISH ; KIAA0418; SH3MD1
Host Species Rabbit
Immunogen Recombinant Protein, aa: 833-892
Purification Method Protein A purfied
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) NM_014631.2
Target Species Human, Mouse
Content And Storage 2-8°C for 12 months in undiluted aliquotes
Form Purified
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.