Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TLL1, Mouse, MaxPab™ Polyclonal Antibody, Abnova™

Catalog No. 89024828 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-024-828 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-024-828 Supplier Abnova Corporation Supplier No. H00007092B01
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a full-length human TLL1 protein.

This gene encodes an astacin-like zinc-dependent metalloprotease and is a subfamily member of the metzincin family. A similar protein in mice is required during heart development and specifically processes procollagen C-propeptides and chordin at similar cleavage sites. [provided by RefSeq

Sequence: MGLGTLSPRMLVWLVASGIVFYGELWVCAGLDYDYTFDGNEEDKTETIDYKDPCKAAVFWGDIALDDEDLNIFQIDRTIDLTQNPFGNLGHTTGGLGDHAMSKKRGALYQLIDRIRRIGFGLEQNNTVKGKVPLQFSGQNEKNRVPRAATSRTERIWPGGVIPYVIGGNFTGSQRAMFKQAMRHWEKHTCVTFIERSDEESYIVFTYRPCGCCSYVGRRGNGPQAISIGKNCDKFGIVVHELGHVIGFWHEHTRPDRDNHVTIIRENIQPGQEYNFLKMEPGEVNSLGERYDFDSIMHYARNTFSRGMFLDTILPSRDDNGIRPAIGQRTRLSKGDIAQARKLYRCPACGETLQESNGNLSSPGFPNGYPSYTHCIWRVSVTPGEKVVFSLC

Specifications

Antigen TLL1
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a full-length human TLL1 protein.
Formulation No additive
Gene TLL1
Gene Accession No. BC016922.2
Gene Alias TLL
Gene Symbols TLL1
Host Species Mouse
Immunogen TLL1 (AAH16922.1, 1 a.a. ∼ 392 a.a) full-length human protein.
Purification Method Unpurified
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7092
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.