Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TLR1 Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody has been used in 13 publications

$206.50 - $501.50

Specifications

Antigen TLR1
Concentration 1.0 mg/mL
Dilution Western Blot 1-3 ug/ml, Flow Cytometry 2-4 ug/ 1x10-6 cells, Immunohistochemistry reported in scientific literature (PMID 27008696), Immunohistochemistry-Paraffin 10 ug/ml, Dot Blot, Flow (Cell Surface) reported in scientific literature (Wong et al (2007); PMID 23593278), Flow (Intracellular) 0.5-1 ug/1 million cells
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin), Dot Blot
Classification Polyclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB10056563T
SDP
View Documents
Novus Biologicals
NB10056563SS
0.025 mg
Each for $206.50
Only null left
Add to Cart
 
NB10056563
SDP
View Documents
Novus Biologicals
NB10056563
0.1 mg
Each for $501.50
Only null left
Add to Cart
 
Description

Description

TLR1 Polyclonal specifically detects TLR1 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin, Dot Blot, Flow (Cell Surface), Flow (Intracellular).
Specifications

Specifications

TLR1
Western Blot 1-3 ug/ml, Flow Cytometry 2-4 ug/ 1x10-6 cells, Immunohistochemistry reported in scientific literature (PMID 27008696), Immunohistochemistry-Paraffin 10 ug/ml, Dot Blot, Flow (Cell Surface) reported in scientific literature (Wong et al (2007); PMID 23593278), Flow (Intracellular) 0.5-1 ug/1 million cells
Polyclonal
Purified
RUO
Human, Mouse, Rat
NP_003254
7096
This antibody was developed against a KLH-conjugated synthetic peptide corresponding to a sequence within amino acids 400-450 of human TLR1.
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
1.0 mg/mL
Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin), Dot Blot
Unconjugated
Rabbit
Cytokine Research, Immunology, Innate Immunity, Signal Transduction, Toll Like Receptors
PBS with 0.05% Sodium Azide
CD281, CD281 antigen, DKFZp547I0610, DKFZp564I0682, KIAA0012MGC126311, MGC104956, MGC126312, rsc786, TIL, TIL. LPRS5, Toll/interleukin-1 receptor-like protein, toll-like receptor 1
TLR1
IgG
Protein G purified
In Ramos, an 87 kDa band is observed.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.