Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ anti-TLR4 Clone: 1H7,

Catalog No. AP1179100UG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
AP1179100UG 100 μg Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AP1179100UG Supplier MilliporeSigma™ Supplier No. AP1179100UG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Specifically detects TLR4 Clone: 1H7 in Human samples, and it is validated for ELISA, Immuno Blotting

TLR4 is a member of the Toll-like receptor (TLR) family which plays a fundamental role in pathogen recognition and activation of innate immunity. TLRs are highly conserved from Drosophila to humans and share structural and functional similarities. They recognize pathogen-associated molecular patterns (PAMPs) that are expressed on infectious agents, and mediate the production of cytokines necessary for the development of effective immunity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TLR4
Applications ELISA, Immunoblot, Immunoblot
Classification Monoclonal
Clone 1H7
Conjugate Unconjugated
Formulation In PBS, pH 7.2.
Host Species Mouse
Immunogen A recombinant polypeptide corresponding to amino acids of 214-292 human TLR4, expressed as a GST fusion protein
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Recognizes the TLR4 protein.
Target Species Human
Content And Storage −20°C
Form Purified
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.