Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TLR5 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA141139
Description
TLR5 Polyclonal antibody specifically detects TLR5 in Human, Mouse, Porcine, Rat samples. It is validated for Flow Cytometry, Immunohistochemistry (Paraffin), In vitro Assay, Western Blot.Specifications
TLR5 | |
Polyclonal | |
Unconjugated | |
TLR5 | |
FLJ10052; MELIOS; MGC126430; MGC126431; RP11-239E10.1; SLE1; SLEB1; systemic lupus erythematosus susceptibility 1; TIL3; Tlr5; toll like receptor 5; Toll/interleukin-1 receptor-like protein 3; toll-like receptor 5 | |
Rabbit | |
Protein G | |
RUO | |
100144476, 289337, 53791, 7100 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Flow Cytometry, Immunohistochemistry (Paraffin), In vitro Assay, Western Blot | |
1.0 mg/mL | |
PBS with 0.05% sodium azide | |
O60602, Q9JLF7 | |
TLR5 | |
Synthetic peptide, KLH-conjugated, corresponding to residues 300-350 of human TLR5. | |
100 μg | |
Primary | |
Human, Mouse, Porcine, Rat | |
Antibody | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction