Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TLR6 Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody has been used in 1 publication

$181.90 - $402.00

Specifications

Antigen TLR6
Dilution Immunohistochemistry-Paraffin 1: 100
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP224914SS
SDP
View Documents
Novus Biologicals
NBP224914SS
0.025 mL
Each for $181.90
Only null left
Add to Cart
 
NBP224914
SDP
View Documents
Novus Biologicals
NBP224914
0.1 mL
Each for $402.00
Only null left
Add to Cart
 
Description

Description

TLR6 Polyclonal specifically detects TLR6 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications

Specifications

TLR6
Immunohistochemistry (Paraffin)
Unconjugated
Rabbit
Adaptive Immunity, Cytokine Research, Immunology, Innate Immunity, Prostate Cancer, Toll Like Receptors
0.1 ml crude whole Rabbit sera with 50% Glycerol with 0.025% Sodium Azide
10333
Within the range of amino acids 25-56 of human TLR6 protein were used as the immunogen.
Primary
Store at -20C. Avoid freeze-thaw cycles.
Immunohistochemistry-Paraffin 1: 100
Polyclonal
Antisera
RUO
Human
CD286, CD286 antigen, toll-like receptor 6
TLR6
IgG
Unpurified
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.