Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TLR8 Antibody - BSA Free, Novus Biologicals™
SDP

Catalog No. NBP177203 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP177203 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP177203 Supplier Novus Biologicals Supplier No. NBP177203
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

TLR8 Polyclonal specifically detects TLR8 in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen TLR8
Applications Western Blot, ELISA, Immunocytochemistry, Immunofluorescence
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 0.5- 2 ug/ml, ELISA 1:100-1:2000, Immunocytochemistry/Immunofluorescence 2-10 ug/ml
Formulation PBS with 0.02% Sodium Azide
Gene Accession No. NP_619542
Gene Alias CD288, CD288 antigen, MGC119599, MGC119600, toll-like receptor 8
Gene Symbols TLR8
Host Species Rabbit
Immunogen Antibody was raised against a peptide corresponding to 16 amino acids near the middle of human TLR8. The immunogen is located within amino acids 790 - 840 of TLR8. Amino Acid Squence: CASPGDQRGKSIVSLE
Molecular Weight of Antigen 107 kDa
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Cytokine Research, Immunology, Infections (Virus Bacteria and Parasites), Inflammation, Innate Immunity, Signal Transduction, Toll Like Receptors
Primary or Secondary Primary
Gene ID (Entrez) 51311
Target Species Human, Mouse
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.