Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TLR9 Rat anti-Human, PE, Clone: EB72-1665, BD

Catalog No. BDB560425 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB560425 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB560425 Supplier BD Biosciences Supplier No. 560425
Add to Cart
Add to Cart

Rat Monoclonal Antibody

Toll-like receptor 9 (TLR9), also designated as CD289, is a type 1 transmembrane glycoprotein (∽110-119kDa calculated molecular weights for protein isoforms) encoded by the TLR9 gene in chromosome 3. TLR9 is a member of the Toll-like receptor (TLR) family whose members play fundamental roles in pathogen recognition and activation of innate immunity. TLR9 is highly expressed by plasmacytoid dendritic cells and B cells and at lower levels in some other cell types. TLR9 is expressed predominantly in the endoplasmic reticulum. It is recruited to endosomal and lysosomal compartments upon cellular stimulation by bacterial, viral or synthetic DNA molecules that contain unmethylated CpG dinucleotide sequences. TLR9 generated signals lead to the activation of genes that express type-I interferons and proinflammatory cytokines including IL-6 and IL-12. TLR9-mediated signaling is also implicated in acquired immune responses as well as in the pathogenesis of autoimmune diseases. The eB72-1665.1 mAb recognizes the human TLR9 epitope from the peptide sequence, amino acids 273-288.

Host Species: Rat
Clone: eB72-1665
Isotype: IgG2a κ
Species Reactivity: Human

TRUSTED_SUSTAINABILITY

Specifications

Antigen TLR9
Applications Flow Cytometry
Classification Monoclonal
Clone EB72-1665
Conjugate PE
Description CD289, toll-like receptor 9
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Host Species Rat
Purification Method Affinity Purified
Quantity 100 Tests
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.