Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TLS Mouse anti-Human, Mouse, Rat, Clone: 1FU-1D2, MilliporeSigma™

Catalog No. 041552MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
04-155-2MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 04-155-2MI Supplier MilliporeSigma Supplier No. 041552
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Use Anti-TLS Antibody, clone 1FU-1D2 (Mouse Monoclonal Antibody) validated in western blot, immunocytochemistry to detect TLS also known as 75 kDa DNA-pairing protein, FUS-CHOP fusion protein.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TLS
Applications Immunocytochemistry, Western Blot
Classification Monoclonal
Clone 1FU-1D2
Conjugate Unconjugated
Formulation Unpurified mouse monoclonal IgG1κ preservative-free ascites.
Gene Accession No. NP_004951.1
Gene Symbols FUS, CHOP, FUS-CHOP, FUS1, POMp75, TLS, TLS/CHOP, hnRNP-P2
Host Species Mouse
Immunogen Linear peptide corresponding to human TLS.
Purification Method Unpurified
Quantity 100 μL
Primary or Secondary Primary
Target Species Human, Mouse, Rat
Content And Storage −20°C in undiluted aliquots for up to 12 months. Avoid repeated freeze/thaw cycles.
Form Ascites
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.