Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TLT-2 (Treml2) Rat anti-Mouse, Brilliant Violet 421, Clone: MIH47, BD

Catalog No. BDB564185 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB564185 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB564185 Supplier BD Biosciences Supplier No. 564185

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Rat Monoclonal Antibody

The MIH47 monoclonal antibody specifically binds to Trem-like transcript 2 protein (TLT-2) which is also known as TREML2 (Triggering receptor expressed on myeloid cells-like 2). TLT-2 is a type I transmembrane protein that belongs to the TREM family. It consists of a single extra-cellular Ig-like V-type domain, a transmembrane segment and a cytoplasmic tail and reportedly serves as a receptor for B7-H3/CD276. It is expressed on most B cells, CD8+ T cells, activated CD4+ T cells, macrophages and granulocytes and at low levels by some NK cells and dendritic cells. TLT-2 can reportedly serve costimulatory and coinhibitory functions in the immune system. TLT-2 expression is upregulated on neutrophils and macrophages in response to inflammatory conditions in vivo. TLT-2 serves as engulfment receptor for apoptotic cells and plays roles in both innate and adaptive immunity.

The antibody was conjugated to BD Horizon™ BV421 which is part of the BD Horizon™ Brilliant Violet™ family of dyes. With an Ex Max of 407-nm and Em Max at 421-nm, BD Horizon™ BV421can be excited by the violet laser and detected in the standard Pacific Blue™ filter set (eg, 450/50-nm filter). BD Horizon™ BV421conjugates are very bright, often exhibiting a 10 fold improvement in brightness compared to Pacific Blue™ conjugates.

TRUSTED_SUSTAINABILITY

Specifications

Antigen TLT-2 (Treml2)
Applications Flow Cytometry
Classification Monoclonal
Clone MIH47
Concentration 0.2mg/mL
Conjugate Brilliant Violet 421
Description B7-H3 receptor; Tlt2; TRML2; Trem-like transcript 2 protein; Gm750
Host Species Rat
Immunogen Mouse TLT-2 Recombinant Protein
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.