Learn More
Description
RNX (HOX11L2, TLX3) belongs to a family of orphan homeobox genes that encode DNA-binding nuclear transcription factors. Members of the HOX11 gene family are characterized by a threonine-47 replacing cytosine in the highly conserved homeodomain (Dear et al., 1993 [PubMed 8099440]).[supplied by OMIM
Specifications
Specifications
| Antigen | T-cell leukemia homeobox 3 |
| Applications | ELISA, Western Blot |
| Classification | Monoclonal |
| Clone | 5B11 |
| Conjugate | Unconjugated |
| Description | Mouse monoclonal antibody raised against a partial recombinant TLX3. |
| Formulation | PBS with no preservative; pH 7.4 |
| Gene | TLX3 |
| Gene Accession No. | NM_021025 |
| Gene Alias | HOX11L2/MGC29804/RNX |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.