Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TMEM106B Rat anti-Human, Rat, Clone: TME-N 6F2, MilliporeSigma™

Catalog No. MABN473MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABN473MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABN473MI Supplier MilliporeSigma Supplier No. MABN473
Add to Cart
Add to Cart

Rat Monoclonal Antibody

Anti-TMEM106B, clone TME-N 6F2 detects levels of TMEM106B proteins and has been published and validated for use in western blotting and immunofluoresence.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TMEM106B
Applications Immunofluorescence, Western Blot
Classification Monoclonal
Clone TME-N 6F2
Conjugate Unconjugated
Formulation Purified rat monoclonal IgG2c in buffer containing 0.1MTris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Accession No. NP_001127704
Gene Symbols TMEM106B
Host Species Rat
Immunogen Recombinant protein corresponding to the N-terminus of human TMEM106B.
Purification Method Protein G purified
Quantity 100 μg
Primary or Secondary Primary
Target Species Human, Rat
Content And Storage 2-8°C for one year
Form Purified
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.